4-Benzoylbenzoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

10572-0050 10572-0250
TS10572-0050EA 83 USD
TS10572-0050 TS10572-0250
4-Benzoylbenzoic acid 99%
4-Benzoylbenzoic acid
Formula: C₁₄H₁₀O₃
Melting Pt: 197…200 °C
Storage Temperature: Ambient
MDL Number: MFCD00002560
CAS Number: 611-95-0
EINECS: 210-286-0

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 197 to 200 °C
Titration with NaOH ≥98.5 %
UV at 375 nm A: ≤0.15
Viscosity 0.22 to 0.27 (1 % at 37 °C)
Ash ≤2.0 %
Average molecular weight 60000 to 90000
pH 4.5 to 7.0 (6 % in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR