Benzophenone hydrazone 98+%

Supplier: Thermo Scientific Chemicals

10557-0050 10557-1000 10557-5000
TS10557-0050EA 21 USD
TS10557-0050 TS10557-1000 TS10557-5000
Benzophenone hydrazone 98+%
Benzophenone hydrazone
Formula: C₁₃H₁₂N₂
MW: 196.25 g/mol
Boiling Pt: 225…230 °C (55 mmHg)
Melting Pt: 95…99 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007624
CAS Number: 5350-57-2
EINECS: 226-321-8

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder or needles
Infrared spectrum Conforms
Melting point 95 to 99 °C
Water ≤0.5 %
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR