p-Anisidine 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Anisidine, 4-Aminoanisole, 4-Methoxyaniline

10483-0010 10483-0050 10483-2500
TS10483-0010EA 154 USD
TS10483-0010 TS10483-0050 TS10483-2500
p-Anisidine 99%
p-Anisidine
Formula: C₇H₉NO
MW: 123.15 g/mol
Boiling Pt: 243 °C (1013 hPa)
Melting Pt: 56…59 °C
Density: 1.06 g/cm³
Flash Pt: 122 °C
Storage Temperature: Ambient
MDL Number: MFCD00007864
CAS Number: 104-94-9
EINECS: 203-254-2
UN: 2811
ADR: 6.1,III
Merck Index: 13,00670

Order Now

Specification Test Results

Appearance Grey to brown Crystalline powder or granules or flakes
Infrared spectrum Conforms
Melting point 56 to 59 °C
GC ≥98.5 %
Stabilizer 10 ppm Kerobit(N,N'-Di-sec-butyl-p-phenylenediamine)
Pyrrolidone ≤0.2 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR