m-Anisidine 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Aminoanisole, 3-Methoxyaniline, 3-Anisidine

10482-0010 10482-0050 10482-1000 10482-2500
TS10482-0010EA 572 USD
TS10482-0010 TS10482-0050 TS10482-1000 TS10482-2500
m-Anisidine 99%
m-Anisidine
Formula: C₇H₉NO
MW: 123.15 g/mol
Boiling Pt: 250…251 °C (760 mmHg)
Density: 1.102 g/cm³
Flash Pt: 126 °C
Storage Temperature: Ambient
MDL Number: MFCD00007783
CAS Number: 536-90-3
EINECS: 208-651-4
UN: 2431
ADR: 6.1,III
Merck Index: 13,00670

Order Now

Specification Test Results

Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5800 to 1.5820 (20°C, 589 nm)
GC ≥98.5 %
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR