o-Anisidine 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Anisidine, 2-Aminoanisole, 2-Methoxyaniline

10481-0010 10481-0050 10481-2500
TS10481-0010EA 148 USD
TS10481-0010 TS10481-0050 TS10481-2500
o-Anisidine 99+%
o-Anisidine
Formula: C₇H₉NO
MW: 123.15 g/mol
Boiling Pt: 225 °C (1013 hPa)
Melting Pt: 3…6 °C
Density: 1.092 g/cm³
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00007688
CAS Number: 90-04-0
EINECS: 201-963-1
UN: 2431
ADR: 6.1,III
Merck Index: 13,00670

Order Now

Specification Test Results

Appearance Clear colorless to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.5730 to 1.5755 (20°C, 589 nm)
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR