4-Aminothiophenol 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Aminobenzenethiol, 4-Aminophenyl mercaptan, 4-Mercaptoaniline

10468-0050 10468-0250
TS10468-0050EA 52 USD
TS10468-0050 TS10468-0250
4-Aminothiophenol 96%
4-Aminothiophenol
Formula: C₆H₇NS
MW: 125.19 g/mol
Boiling Pt: 140…145 °C (16 mmHg)
Melting Pt: 37…43 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007880
CAS Number: 1193-02-8
EINECS: 214-763-4
UN: 3263
ADR: 8,III

Order Now

Specification Test Results

Appearance Yellow Low melting crystalline mass
Infrared spectrum Conforms
Melting point 37 to 43 °C
Titration Argentometric ≥95 %
Additional info oxidized (disulphide) compound is steadily formed in air

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR