4-Aminophenol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: p-Aminophenol, UrsolP Rodinal 4-Amino-1-hydroxybenzene p-Hyddroxyaniline 4-Aminophenol 4-Hydroxyaniline, 4-Hydroxyaniline

10427-0010 10427-0050 10427-0051 10427-2500
TS10427-0010EA 119 USD
TS10427-0010 TS10427-0050 TS10427-0051 TS10427-2500
4-Aminophenol 97%
4-Aminophenol
Formula: H₂NC₆H₄OH
MW: 109.13 g/mol
Boiling Pt: 284 °C (760 mmHg)
Melting Pt: 187…191 °C
Flash Pt: 189 °C
Storage Temperature: Ambient
MDL Number: MFCD00007869
CAS Number: 123-30-8
EINECS: 204-616-2
UN: 2512
ADR: 6.1,III
Merck Index: 13,00461
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7af17b754d4ebaa0993b4544d4e6ab6c116724c&vid=6275667c2624ee686f61715acd420dc596e076fa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to cream to beige to light brown Powder or crystals
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR