2-Aminophenol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Amino-1-hydroxy benzene, 2-Hydroxy aniline, 2-Hydroxyaniline, o-Aminophenol

10426-0050 10426-1000 10426-5000
TS10426-0050EA 35 USD
TS10426-0050 TS10426-1000 TS10426-5000
2-Aminophenol 99%
2-Aminophenol
Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 164 °C (15mbar)
Melting Pt: 172…177 °C
Flash Pt: 168 °C
MDL Number: MFCD00007690
CAS Number: 95-55-6
EINECS: 202-431-1
UN: 2512
ADR: 6.1,III
Merck Index: 13,00460

Order Now

Specification Test Results

Appearance (Color) White to brown
Appearance (Form) Powder
Infrared spectrum Conforms
Melting point 172 to 177 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR