2,3-Dihydro-1H-inden-2-amine hydrochloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Aminoindan hydrochloride, 2-Indanylamine hydrochloride
10386-0010
10386-0050
10386-0250
TS10386-0010EA
40
USD
TS10386-0010
TS10386-0050
TS10386-0250
2,3-Dihydro-1H-inden-2-amine hydrochloride 98%
2,3-Dihydro-1H-inden-2-amine hydrochloride
Formula:
C₉H₁₂ClN MW: 169.65 g/mol Melting Pt: 246…247 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012549 CAS Number: 2338-18-3 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
GC | ≥90 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...