(±)-1-Aminoindan 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Aminoindan, 1-Indanamine, 2,3-Dihydro-1H-inden-1-amine, (±)-1-Indanylamine

10385-0010 10385-0050
TS10385-0010EA 74 USD
TS10385-0010 TS10385-0050
(±)-1-Aminoindan 98%
(±)-1-Aminoindan
Formula: C₉H₁₁N
MW: 133.19 g/mol
Boiling Pt: 96…97 °C (8 mmHg)
Melting Pt: 1.5 °C
Density: 1.038 g/cm³ (20 °C)
Flash Pt: 94 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003799
CAS Number: 34698-41-4
EINECS: 252-158-7

Order Now

Specification Test Results

Appearance Clear colorless to light yellow to green Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5600 to 1.5640 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR