4-Amino-2,6-dichlorophenol 98%

Supplier: Thermo Scientific Chemicals
Warning

10348-0050 10348-0250
TS10348-0050EA 21 USD
TS10348-0050 TS10348-0250
4-Amino-2,6-dichlorophenol 98%
4-Amino-2,6-dichlorophenol
Formula: C₆H₅Cl₂NO
MW: 178.02 g/mol
Boiling Pt: 304 °C (1013 hPa)
Melting Pt: 164…168 °C
MDL Number: MFCD00007875
CAS Number: 5930-28-9
EINECS: 227-671-4

Order Now

Specification Test Results

Appearance Beige to light brown to light red Powder
Infrared spectrum Conforms
Melting point 164 to 168 °C
Solubility (2.5 % in 96 % ethanol) Clear to slightly hazy
Titration with TBAH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR