2-Amino-5-chlorobenzophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Amino-4-chloro-2-(phenylmethanoyl)benzene, (2-Amino-5-chlorophenyl)(phenyl)methanone

10334-0250 10334-1000 10334-5000
TS10334-0250EA 24 USD
TS10334-0250 TS10334-1000 TS10334-5000
2-Amino-5-chlorobenzophenone 98%
2-Amino-5-chlorobenzophenone
Formula: C₁₃H₁₀ClNO
MW: 231.68 g/mol
Boiling Pt: 207 °C (760 mmHg)
Melting Pt: 96…100 °C
Flash Pt: 211 °C
Storage Temperature: Ambient
MDL Number: MFCD00007839
CAS Number: 719-59-5
EINECS: 211-949-7

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 96 to 100 °C
GC ≥97.5 %
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR