α,α,α-Trifluoro-m-toluidine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-(Trifluoromethyl)aniline, 3-Aminobenzotrifluoride

10319-0050 10319-1000 10319-5000
TS10319-0050EA 26 USD
TS10319-0050 TS10319-1000 TS10319-5000
α,α,α-Trifluoro-m-toluidine 98%
α,α,α-Trifluoro-m-toluidine
Formula: C₇H₆F₃N
MW: 161.13 g/mol
Boiling Pt: 187…193 °C (760 mmHg)
Melting Pt: 5…6 °C
Density: 1.303 g/cm³
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00007797
CAS Number: 98-16-8
EINECS: 202-643-4
UN: 2948
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow to orange Liquid
Infrared spectrum Conforms
Refractive index 1.4790 to 1.4810 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR