α,α,α-Trifluoro-o-toluidine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(Trifluoromethyl)aniline, 2-Aminobenzotrifluoride

10318-0025 10318-0050 10318-1000 10318-5000
TS10318-0025EA 752 USD
TS10318-0025 TS10318-0050 TS10318-1000 TS10318-5000
α,α,α-Trifluoro-o-toluidine 99%
α,α,α-Trifluoro-o-toluidine
Formula: C₇H₆F₃N
MW: 161.13 g/mol
Boiling Pt: 175
Melting Pt: -33
Density: 1.296 g/cm³
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00007718
CAS Number: 88-17-5
EINECS: 201-806-7
UN: 2942
ADR: 6.1 III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4800 to 1.4830 (20°C, 589 nm)
GC ≥98.5 %
Titration excess NaOH 98.0 to 100.5 %
Iron (Fe) ≤0.001 %
Heavy metals (as Pb) ≤10 ppm
Phosphorous compounds (as P) ≤0.002 %
Freezing point -2.0 to 0.0°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR