4-Aminobenzophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (4-Aminophenyl)(phenyl)methanone, p-Aminobenzophenone

10317-0100 10317-0250
TS10317-0100EA 69 USD
TS10317-0100 TS10317-0250
4-Aminobenzophenone 98%
4-Aminobenzophenone
Formula: C₁₃H₁₁NO
MW: 197.24 g/mol
Boiling Pt: 246 °C (17hPa)
Melting Pt: 120…124 °C
Storage Temperature: Ambient
MDL Number: MFCD00007895
CAS Number: 1137-41-3
EINECS: 214-506-6

Order Now

Specification Test Results

Appearance Yellow to yellow-green to beige to light grey to orange to brown Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 120 to 124 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR