2-Aminobenzophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

10316-0250 10316-1000
TS10316-0250EA 90 USD
TS10316-0250 TS10316-1000
2-Aminobenzophenone 98%
2-Aminobenzophenone
Formula: C₁₃H₁₁NO
Boiling Pt: 384 °C (1013 hPa)
Melting Pt: 105…108 °C
Flash Pt: 240 °C
Storage Temperature: Ambient
MDL Number: MFCD00007713
CAS Number: 2835-77-0
EINECS: 220-613-9

Order Now

Specification Test Results

Appearance Yellow Powder and/or chunks
Infrared spectrum Conforms
Melting point 105 to 108 °C
Titration with HClO4 ≥97.5 %
Titration by coupling ≥97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR