9-Acetylanthracene 95%

Supplier: Thermo Scientific Chemicals

10244-0050 10244-0250
TS10244-0050EA 43 USD
TS10244-0050 TS10244-0250
9-Acetylanthracene 95%
9-Acetylanthracene
Formula: C₁₆H₁₂O
Melting Pt: 72…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00001259
CAS Number: 784-04-3
EINECS: 212-315-2

Order Now

Specification Test Results

Appearance Yellow to yellow-brown Crystalline powder
Infrared spectrum Conforms
Melting point 72 to 76 °C
GC ≥94 %
Iron (Fe) ≤0.002 %
Magnesium (Mg) ≤0.002 %
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.02 %
Heavy metals (as Pb) ≤0.001 %
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR