Paracetamol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Acetaminophen, N-Acetyl-4-aminophenol, 4′-Hydroxyacetanilide, 4'-Hydroxyacetanilide, Paracetamolum, 4-Acetamidophenol, N-(4-Hydroxyphenyl)acetamide, APAP

10233-0010 10233-0050 10233-0051 10233-2500
TS10233-0010EA 163 USD
TS10233-0010 TS10233-0050 TS10233-0051 TS10233-2500
Paracetamol 98%
Paracetamol
Formula: C₈H₉NO₂
MW: 151.16 g/mol
Melting Pt: 168…172 °C
Density: 1.293 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00002328
CAS Number: 103-90-2
EINECS: 203-157-5
Merck Index: 13,00048

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 168 to 172 °C
HPLC ≥97.5 %
Gel strength 500 to 1000 g/cm² (1.5 % at 20 °C)
Ash ≤6 %
Microbial contamination Absent (Salmonella)
pH 5.7 to 7.5 (1.5 % in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR