2'-Hydroxyacetanilide 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-Acetyl-2-aminophenol, N-(2-Hydroxyphenyl)acetamide, 2-Acetamidophenol, Acetylcholine iodide

10231-0250 10231-1000
TS10231-0250EA 28 USD
TS10231-0250 TS10231-1000
2'-Hydroxyacetanilide 97%
2'-Hydroxyacetanilide
Formula: C₈H₉NO₂
MW: 151.16 g/mol
Melting Pt: 207…210 °C
MDL Number: MFCD00002181
CAS Number: 614-80-2
EINECS: 210-396-9

Order Now

Specification Test Results

Appearance Off-white to beige to brown Powder
Infrared spectrum Conforms
Melting point 205 to 210 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR