Mebendazole

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl [5-(Benzoyl)benzimidazol-2-yl]carbamate, Methyl (5-benzoyl-1H-benzo[d]imidazol-2-yl)carbamate

10172-0050 10172-0250
TS10172-0050EA 36 USD
TS10172-0050 TS10172-0250
Mebendazole
Mebendazole
Formula: C₁₆H₁₃N₃O₃
MW: 295.30 g/mol
Melting Pt: 287 °C
Storage Temperature: Ambient
MDL Number: MFCD00057872
CAS Number: 31431-39-7

Order Now

Specification Test Results

Appearance White to off-white to grey Powder
Infrared spectrum Conforms
Melting point 285 to 290 °C
Loss on drying ≤0.5 % (1 g, 105 °C, 4 h)
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR