2-Vinylanthraquinone ≥98.0% (by GC)
Supplier: TCI America
V0139-1G
V0139-5G
TCV0139-1GEA
133.93
USD
TCV0139-1G
TCV0139-5G
2-Vinylanthraquinone ≥98.0% (by GC)
2-Vinylanthraquinone
Formula:
C₁₆H₁₀O₂ Melting Pt: 177°C |
CAS Number:
13388-33-5 |
Specification Test Results
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...