Vac Elut SPS 24 Manifold, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
12234003 12234004 12234022 12234041
AG12234003EA 3999.78 USD
AG12234003 AG12234004 AG12234022 AG12234041
Vac Elut SPS 24 Manifold, Agilent Technologies
Manifolds
Vac Elut 24 SPS manifold processes up to 24 SPE cartridges at the same time.
  • Closed operation prevents cross contamination
  • Stainless steel tips deliver maximum extract purity
  • Range of rack sizes covers most tube configurations
The Vac Elut SPS 24 manifold allows you to process up to 24 SPE cartridges simultaneously. Like all Vac Elut manifolds, the SPS 24 is made from durable, solvent-resistant materials and engineered to last. The glass sides allow easy viewing of the entire sample collection process.