SPME Fiber Accessories, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
5191-5877 392609902
AG5191-5877KT 1330 USD
AG5191-5877 AG392609902
SPME Fiber Accessories, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Extra parts to assist in SPME sample preparation.
  • SPME fiber kits contain only the fibers
For a first-time order, you will also need to order the appropriate fiber holder for your application needs.