Mopec Swordfish Saw and Vacuum Set, Mortech
Supplier: Mortech Manufacturing
BD090+BD093 BD094+BD095
76549-618EA 8833.4 USD
76549-618 76549-620
Mopec Swordfish Saw and Vacuum Set, Mortech
Saws
Previously known as the Mopec 5000, the Mopec Swordfish is the first bone saw developed specifically for autopsy use.
  • Reduced vibration
It is totally submersible for complete cleaning. The hand-piece runs on 40V for a cooler touch, reduced vibration; and the axial mount blades offer safer installation. Compatible with the Mopec Swordfish Autopsy Saw only, the BD093 vacuum will extract harmful debris and matter from the air. Flexible, clear plastic hood improves visibility.
Información para pedidos: For available accessories, contact your Avantor representative.