DSS (Disuccinimidyl suberate)

Supplier: G-Biosciences

Synonyms: Bis(2,5-dioxopyrrolidin-1-yl) octanedioate, Suberic acid bis(N-hydroxysuccinimide ester), Disuccinimidyl octanedioate, DSS

BC04 BC04-Q
82022-776EA 403.8 USD
82022-776 76286-304
DSS (Disuccinimidyl suberate)
DSS (Disuccinimidyl suberate)

Disuccinimidyl suberate Homobifunctional Cross Linker.


  • Ideal for receptor ligand cross-linking and coupling antibody to Protein A or G for immunoprecipitations.
  • OneQuant™ DSS is supplied as 8 vials of 2mg DSS, suitable for preparing for ~100µl 50mM stock solution from each vial.

Formula: C₁₆H₂₀N₂O₈
MW: 368.34 g/mol
MDL Number: MFCD00049059
CAS Number: 68528-80-3

Order Now

Specification Test Results

Spacer Arm (Å) 11.4
Reactive Toward Primary Amines
Membrane Permeable Yes
Water Soluble No
Cleavable/ Reversible No


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR