Dibutyl carbonate

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Carbonic acid dibutyl ester

46388-0050 46388-0250
TS46388-0050EA 55 USD
TS46388-0050 TS46388-0250
Dibutyl carbonate
Dibutyl carbonate
Formula: C₉H₁₈O₃
MW: 174.24 g/mol
Density: 0.92
Storage Temperature: Ambient
MDL Number: MFCD00043938
CAS Number: 542-52-9

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4100 to 1.4140 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR