Desvenlafaxine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(2-(Dimethylamino)-1-(1-hydroxycyclohexyl)-ethyl)phenol, O-Desmethylvenlafaxine, 4-[2-(Dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]phenol, Desvenlafaxine

46157-0010 46157-0050
TS46157-0010EA 60 USD
TS46157-0010 TS46157-0050
Desvenlafaxine 97%
Desvenlafaxine
Formula: C₁₆H₂₅NO₂
MW: 263.38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00871934
CAS Number: 93413-62-8

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR