5-Hydroxy-4-octanone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Hydroxyoctan-4-one, 5-Hydroxy-4-octanone

46076-0050 46076-0250
TS46076-0050EA 82 USD
TS46076-0050 TS46076-0250
5-Hydroxy-4-octanone 97%
5-Hydroxy-4-octanone
Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 188
Melting Pt: 2
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00021928
CAS Number: 496-77-5
Merck Index: 13,01595

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR