Morpholin-3-one 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
46053-0010
46053-0050
46053-0250
TS46053-0010EA
32
USD
TS46053-0010
TS46053-0050
TS46053-0250
Morpholin-3-one 98%
Morpholin-3-one
Formula:
C₄H₇NO₂ Melting Pt: 106…108 °C Storage Temperature: Ambient |
MDL Number:
MFCD00631009 CAS Number: 109-11-5 |
Specification Test Results
Appearance | Colorless to white Crystalline solid |
Infrared spectrum | Conforms |
Melting point | 102 to 109 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...