3,5-Di-tert-butyl-2-hydroxybenzaldehyde 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
45812-0050
45812-0250
TS45812-0050EA
58
USD
TS45812-0050
TS45812-0250
3,5-Di-tert-butyl-2-hydroxybenzaldehyde 99%
3,5-Di-tert-butyl-2-hydroxybenzaldehyde
Formula:
C₁₅H₂₂O₂ Melting Pt: 61…62 °C Storage Temperature: Ambient |
MDL Number:
MFCD00191998 CAS Number: 37942-07-7 |
Specification Test Results
Appearance | White to yellow Powder or crystalline powder |
Infrared spectrum | Conforms |
Melting point | 56 to 63 °C |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...