Decyl decanoate 98%

Supplier: Thermo Scientific Chemicals

45711-0250 45711-5000
TS45711-0250EA 47 USD
TS45711-0250 TS45711-5000
Decyl decanoate 98%
Decyl decanoate
Formula: C₂₀H₄₀O₂
Boiling Pt: 174 °C (2 mmHg)
Density: 0.859 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00059280
CAS Number: 1654-86-0

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4410 to 1.4430 (20 °C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR