3-(Bromoacetyl)pyridine ydrobromide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(2-Bromoacetyl)pyridine hydrobromide

45552-0050 45552-0250
TS45552-0050EA 45 USD
TS45552-0050 TS45552-0250
3-(Bromoacetyl)pyridine ydrobromide 98%
3-(Bromoacetyl)pyridine ydrobromide
Formula: C₇H₆BrNO
MW: 280.95 g/mol
Melting Pt: 192…196 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00052182
CAS Number: 17694-68-7

Order Now

Specification Test Results

Appearance White to yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 188 to 197 °C
Titration Argentometric ≥97.5 %
Water ≤0.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR