6-Bromoisatin 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Bromo-1H-indole-2,3-dione, 6-Bromoindoline-2,3-dione

45551-0050 45551-0250
TS45551-0050EA 77 USD
TS45551-0050 TS45551-0250
6-Bromoisatin 95%
6-Bromoisatin
Formula: C₈H₄BrNO₂
MW: 226.03 g/mol
Melting Pt: 274 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01631138
CAS Number: 6326-79-0

Order Now

Specification Test Results

Appearance Yellow to orange Powder
Infrared spectrum Conforms
Melting point 267 to 275 °C
Loss on drying ≤3 %
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR