3-Methoxypropionic acid 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Methoxypropanoic acid

45332-0100 45332-0500
TS45332-0100EA 163 USD
TS45332-0100 TS45332-0500
3-Methoxypropionic acid 97+%
3-Methoxypropionic acid
Formula: C₄H₈O₃
MW: 104.1100 g/mol
Boiling Pt: 116 °C (9 mmHg)
Density: 1.108 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00042654
CAS Number: 2544-06-1

Order Now

Specification Test Results

Appearance Colorless Liquid
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR