2,4-Diacetylphloroglucinol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,1'-(2,4,6-Trihydroxy-1,3-phenylene)bisethanone, 1-(3-acetyl-2,4,6-trihydroxyphenyl)ethan-1-one, Ethanone,1,1'-(2,4,6-trihydroxy-1,3-phenylene)bis-

45280-0010 45280-0050
TS45280-0010EA 237 USD
TS45280-0010 TS45280-0050
2,4-Diacetylphloroglucinol 97%
2,4-Diacetylphloroglucinol
Formula: C₁₀H₁₀O₅
MW: 210.19 g/mol
Melting Pt: 164…169 °C
Storage Temperature: Ambient
MDL Number: MFCD00100483
CAS Number: 2161-86-6

Order Now

Specification Test Results

Appearance Off-white to light brown Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR