3,5-Diiodosalicylaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

45086-0050 45086-0250
TS45086-0050EA 51 USD
TS45086-0050 TS45086-0250
3,5-Diiodosalicylaldehyde 97%
3,5-Diiodosalicylaldehyde
Formula: C₇H₄I₂O₂
Melting Pt: 108…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003321
CAS Number: 2631-77-8

Order Now

Specification Test Results

Appearance Yellow to light brown Powder or solid
Infrared spectrum Conforms
Melting point 107 to 112 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR