2,6-Dichlorophenolindophenol sodium salt hydrate 90+%
Supplier: Thermo Scientific Chemicals
Synonyms:
DPIP, DCIP
45070-0050
45070-0250
45070-1000
TS45070-0050EA
55
USD
TS45070-0050
TS45070-0250
TS45070-1000
2,6-Dichlorophenolindophenol sodium salt hydrate 90+%
2,6-Dichlorophenolindophenol sodium salt hydrate
Formula:
C₁₂H₆Cl₂NNaO₂·xH₂O MW: 290.08 g/mol (anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00150014 CAS Number: 1266615-56-8 EINECS: 210-640-4 |
Specification Test Results
Appearance | Dark green to dark blue-purple Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤14 % (120 °C) |
Titration with HClO4 | ≥90.0 % (On dry substance) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...