2-Fluoro-5-formylpyridine 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
6-Fluoropyridine-3-carboxaldehyde, 6-Fluoronicotinaldehyde
44985-0010
44985-0050
44985-0250
TS44985-0010EA
61
USD
TS44985-0010
TS44985-0050
TS44985-0250
2-Fluoro-5-formylpyridine 95%
2-Fluoro-5-formylpyridine
Formula:
C₆H₄FNO MW: 125.1 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08064049 CAS Number: 677728-92-6 |
Specification Test Results
Appearance | Colorless to white to light yellow Liquid or low melting solid |
Infrared spectrum | Conforms |
GC | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...