4-Bromo-2-furaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Bromofurfural, 4-Bromofuran-2-carbaldehyde

44944-0010 44944-0050
TS44944-0010EA 102 USD
TS44944-0010 TS44944-0050
4-Bromo-2-furaldehyde 97%
4-Bromo-2-furaldehyde
Formula: C₅H₃BrO₂
MW: 174.98 g/mol
Melting Pt: 53…54 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00671504
CAS Number: 21921-76-6

Order Now

Specification Test Results

Appearance White to brown Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 53 to 57 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR