5-Methoxyisatin 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Methoxy-1H-indole-2,3-dione, 5-Methoxyindoline-2,3-dione

44883-0050 44883-0250 44883-1000
TS44883-0050EA 57 USD
TS44883-0050 TS44883-0250 TS44883-1000
5-Methoxyisatin 97%
5-Methoxyisatin
Formula: C₉H₇NO₃
MW: 177.16 g/mol
Melting Pt: 201…203 °C
Storage Temperature: Ambient
MDL Number: MFCD00169023
CAS Number: 39755-95-8

Order Now

Specification Test Results

Appearance Dark red to brown Powder
Infrared spectrum Conforms
Melting point 199 to 202 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR