ω-Bromo-2-fluoroacetophenone 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Bromomethyl o-fluorophenyl ketone, 2-Bromo-2'-fluoroacetophenone, α-Bromo-2-fluoroacetophenone, 2-Fluorophenacyl bromide, 2-Bromo-1-(2-fluorophenyl)ethan-1-one, 2-Bromo-1-(2-fluorophenyl)ethanone

44667-0050 44667-0250
TS44667-0050EA 102 USD
TS44667-0050 TS44667-0250
ω-Bromo-2-fluoroacetophenone 98%
ω-Bromo-2-fluoroacetophenone
Formula: C₈H₆BrFO
MW: 217.04 g/mol
Boiling Pt: 61…63 °C (.1Torr)
Storage Temperature: Freezer
MDL Number: MFCD00278796
CAS Number: 655-15-2
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Appearance Yellow to green to brown Viscous liquid or low melting solid
Infrared spectrum Conforms
GC ≥97.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR