2,4-Dimethoxy-5-pyrimidineboronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,4-Dimethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine, 2,4-Dimethoxypyrimidine-5-boronic acid pinacol ester

44493-0010 44493-0050
TS44493-0010EA 171 USD
TS44493-0010 TS44493-0050
2,4-Dimethoxy-5-pyrimidineboronic acid pinacol ester 97%
2,4-Dimethoxy-5-pyrimidineboronic acid pinacol ester
Formula: C₁₂H₁₉BN₂O₄
MW: 266.1013 g/mol
Melting Pt: 104…109 °C
Storage Temperature: Ambient
MDL Number: MFCD06795654
CAS Number: 936250-17-8

Order Now

Specification Test Results

Appearance White to off-white Solid
Infrared spectrum Conforms
Melting point 104 to 109 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR