3-Bromo-5-formylpyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

44142-0010 44142-0050
TS44142-0010EA 52 USD
TS44142-0010 TS44142-0050
3-Bromo-5-formylpyridine 97%
3-Bromo-5-formylpyridine
Formula: C₆H₄BrNO
Melting Pt: 96…98 °C
Storage Temperature: Ambient
MDL Number: MFCD03265758
CAS Number: 113118-81-3

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 95 to 99 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR