2-Chloropyrimidine-5-carboxyaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Chloropyrimidine-5-carboxaldehyde, 2-Chloropyrimidine-5-carbaldehyde

43845-0010 43845-0050
TS43845-0010EA 260 USD
TS43845-0010 TS43845-0050
2-Chloropyrimidine-5-carboxyaldehyde 97%
2-Chloropyrimidine-5-carboxyaldehyde
Formula: C₅H₃ClN₂O
MW: 142.55 g/mol
Melting Pt: 113…118 °C
Storage Temperature: Ambient
MDL Number: MFCD10696891
CAS Number: 933702-55-7

Order Now

Specification Test Results

Appearance White to yellow Powder
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR