3-Methyl-3-penten-2-one 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Methylpent-3-en-2-one

43844-0100 43844-0500
TS43844-0100EA 85 USD
TS43844-0100 TS43844-0500
3-Methyl-3-penten-2-one 98%
3-Methyl-3-penten-2-one
Formula: C₆H₁₀O
MW: 98.14 g/mol
Boiling Pt: 136…139 °C
Melting Pt: –70 °C
Density: 0.875 g/cm³
Flash Pt: 34 °C
Storage Temperature: Ambient
MDL Number: MFCD00151839
CAS Number: 565-62-8
UN: 1992
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR