4,4,5,5,5-Pentafluoropentan-1-ol 95%

Supplier: Thermo Scientific Chemicals
Warning

43843-0050 43843-0250
TS43843-0050EA 106 USD
TS43843-0050 TS43843-0250
4,4,5,5,5-Pentafluoropentan-1-ol 95%
4,4,5,5,5-Pentafluoropentan-1-ol
Formula: C₅H₇F₅O
MW: 178.1 g/mol
Boiling Pt: 134…135 °C
Flash Pt: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD00153224
CAS Number: 148043-73-6

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR