Methyl tridecanoate 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Tridecylic acid methyl ester, Tridecanoic acid methyl ester

43820-0010 43820-0050 43820-0100
TS43820-0010EA 49 USD
TS43820-0010 TS43820-0050 TS43820-0100
Methyl tridecanoate 97%
Methyl tridecanoate
Formula: C₁₄H₂₈O₂
Boiling Pt: 130…132 °C (4Torr)
Melting Pt: 7 °C
Density: 0.86 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008977
CAS Number: 1731-88-0

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR