5-Acetyl-2-methoxypyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

43817-0010 43817-0050
TS43817-0010EA 65 USD
TS43817-0010 TS43817-0050
5-Acetyl-2-methoxypyridine 97%
5-Acetyl-2-methoxypyridine
Formula: C₈H₉NO₂
MW: 151.1600 g/mol
Melting Pt: 53…54 °C
Storage Temperature: Ambient
MDL Number: MFCD05664350
CAS Number: 213193-32-9

Order Now

Specification Test Results

Appearance White to yellow Powder
Infrared spectrum Conforms
Melting point 52.0 to 57.0 °C
Water ≤3.00 %
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR