(R)-(+)-3-Chloro-1-phenyl-1-propanol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (R)-3-Chloro-1-phenylpropan-1-ol

43204-0010 43204-0050
TS43204-0010EA 54 USD
TS43204-0010 TS43204-0050
(R)-(+)-3-Chloro-1-phenyl-1-propanol 97%
(R)-(+)-3-Chloro-1-phenyl-1-propanol
Formula: C₉H₁₁ClO
Melting Pt: 59.5…59.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00075128
CAS Number: 100306-33-0

Order Now

Specification Test Results

Appearance White to yellow Powder
Infrared spectrum Conforms
Melting point 58 to 61 °C
GC ≥96.0 %
Specific optical rotation +23° to +29° (20 °C, 589 nm) (c=1 in Chloroform)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR