1,3-Benzothiazole-6-carboxaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Benzothiazolecarboxaldehyde, Benzo[d]thiazole-6-carbaldehyde, Benzothiazol-6-carboxaldehyde, 6-Formylbenzothiazole

43115-0010 43115-0050
TS43115-0010EA 267 USD
TS43115-0010 TS43115-0050
1,3-Benzothiazole-6-carboxaldehyde 97%
1,3-Benzothiazole-6-carboxaldehyde
Formula: C₈H₅NOS
Melting Pt: 92…93 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03086101
CAS Number: 19989-67-4

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 90 to 96 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR